Syklobutankarboksyl syre ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
C0888-10G
C0888-25G
TCIAC0888-10GEA
485
NOK
TCIAC0888-10G
TCIAC0888-25G
Syklobutankarboksyl syre ≥97.0% (by GC, titration analysis)
Syklobutankarboksyl syre
Formel:
C₅H₈O₂ MW: 100,12 g/mol Kokepunkt: 196°C Smeltepunkt: –20 °C Tetthet: 1,06 g/cm³ (20 °C) Flash punkt: 83°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00001323 CAS-nummer: 3721-95-7 EINECS: 223-072-7 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...