Syklopentankarboksyl syre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

C0512-25G C0512-5G
TCIAC0512-25GEA 610 NOK
TCIAC0512-25G TCIAC0512-5G
Syklopentankarboksyl syre ≥98.0% (by GC, titration analysis)
Syklopentankarboksyl syre
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokepunkt: 216°C
Smeltepunkt: 4 °C
Tetthet: 1,05 g/cm³ (20 °C)
Flash punkt: 94°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001371
CAS-nummer: 3400-45-1
EINECS: 222-269-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd651b2a3a5616f6230c76a47c8028145218a28e&vid=a9b8b3c92b24092fb9a4509a4f3d6639632d7dff&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR