Dimethyl-2-nitroterephthalate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: Dimethyl 2-nitroterephthalate

N0501-25G N0501-250G
TCIAN0501-25GEA 220 NOK
TCIAN0501-25G TCIAN0501-250G
Dimethyl-2-nitroterephthalate ≥97.0% (by GC)
Dimethyl-2-nitroterephthalate
Formel: C₁₀H₉NO₆
MW: 239,18 g/mol
MDL nummer: MFCD00008422
CAS-nummer: 5292-45-5
EINECS: 226-134-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ded79d7a083a728097b96daf1cadb36356bc776&vid=2ded79d7a083a728097b96daf1cadb36356bc776&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 74.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR