Dibutyltin dilaurate ≥95.0% (W)
Leverandør: TCI
|
Danger
|
Synonymer:
Di-n-butyltin dilaurate
D0303-25G
D0303-500G
D0303-100G
TCIAD0303-25GEA
245
NOK
TCIAD0303-25G
TCIAD0303-500G
TCIAD0303-100G
Dibutyltin dilaurate ≥95.0% (W)
Dibutyltin dilaurate
Formel:
C₃₂H₆₄O₄Sn MW: 631,57 g/mol Flash punkt: 236°C |
MDL nummer:
MFCD00008963 CAS-nummer: 77-58-7 EINECS: 201-039-8 FN: 2788 ADR: 6.1,II Merck Indeks: 14,03038 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Gravimetric Method) | min. 95.0 % |
Ignition residue (as SnO2) | 23.4 to 24.6 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...