Ethyl-3,5-dichloro-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Ethyl 3,5-dichloro-4-hydroxybenzoate
D0374-25G
D0374-5G
TCIAD0374-25GEA
2375
NOK
TCIAD0374-25G
TCIAD0374-5G
Ethyl-3,5-dichloro-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Ethyl-3,5-dichloro-4-hydroxybenzoate
Formel:
C₉H₈Cl₂O₃ MW: 235,07 g/mol |
MDL nummer:
MFCD00016420 CAS-nummer: 17302-82-8 EINECS: 241-331-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 110.0 to 115.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...