Tropinone ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
8-Methyl-8-azabicyclo[3.2.1]octan-3-one
T0626-25G
T0626-5G
TCIAT0626-25GEA
1050
NOK
TCIAT0626-25G
TCIAT0626-5G
Tropinone ≥98.0% (by GC, titration analysis)
Tropinone
Formel:
C₈H₁₃NO MW: 139,2 g/mol Kokepunkt: 218°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00005549 CAS-nummer: 532-24-1 EINECS: 208-530-6 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 41.0 to 44.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...