2-Amino-3,5-dichloro-6-methylpyridine ≥97.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Amino-3,5-dichloro-2-methylpyridine
A1933-1G
A1933-5G
TCIAA1933-1GEA
325
NOK
TCIAA1933-1G
TCIAA1933-5G
2-Amino-3,5-dichloro-6-methylpyridine ≥97.0% (by titrimetric analysis)
2-Amino-3,5-dichloro-6-methylpyridine
Formel:
C₆H₆Cl₂N₂ MW: 177,03 g/mol |
MDL nummer:
MFCD00129029 CAS-nummer: 22137-52-6 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...