3,3,5-Trimetylsykloheksanon ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
T0602-25ML
T0602-500ML
TCIAT0602-25MLEA
230
NOK
TCIAT0602-25ML
TCIAT0602-500ML
3,3,5-Trimetylsykloheksanon ≥98.0% (by GC)
3,3,5-Trimetylsykloheksanon
Formel:
C₉H₁₆O MW: 140,23 g/mol Kokepunkt: 190°C Smeltepunkt: –10 °C Tetthet: 0,89 g/cm³ (20 °C) Flash punkt: 68°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00019466 CAS-nummer: 873-94-9 EINECS: 212-855-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...