Dimedon ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione
D0701-25G
D0701-500G
D0701-100G
TCIAD0701-25GEA
325
NOK
TCIAD0701-25G
TCIAD0701-500G
TCIAD0701-100G
Dimedon ≥99.0% (by titrimetric analysis)
Dimedon
Formel:
C₈H₁₂O₂ MW: 140,18 g/mol Kokepunkt: 234 °C (1013 hPa) Smeltepunkt: 148 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00001588 CAS-nummer: 126-81-8 EINECS: 204-804-4 Merck Indeks: 14,03242 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 147.0 to 151.0 deg-C |
Solubility in hot Water | almost transparency |
Ignition residue (Sulfate) | max. 0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...