5-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 5-Methylcyclohexane-1,3-dione
M0945-1G M0945-5G
TCIAM0945-1GEA 300 NOK
TCIAM0945-1G TCIAM0945-5G
5-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
5-Methyl-1,3-cyclohexanedione
Formel: C₇H₁₀O₂
MW: 126,16 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010379
CAS-nummer: 4341-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d453b25ae80857d959d4aca41ad0f2ac9f967f1c&vid=d453b25ae80857d959d4aca41ad0f2ac9f967f1c&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 129.0 deg-C
Solubility in Methanol almost transparency