6-Hydroxypicolinic acid ≥97.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid
H0876-25G
H0876-5G
TCIAH0876-25GEA
2325
NOK
TCIAH0876-25G
TCIAH0876-5G
6-Hydroxypicolinic acid ≥97.0% (by titrimetric analysis)
6-Hydroxypicolinic acid
Formel:
C₆H₅NO₃ MW: 139,11 g/mol |
MDL nummer:
MFCD00192220 CAS-nummer: 19621-92-2 EINECS: 417-690-8 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...