5-Bromopyridine-2-carbonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

B3610-25G B3610-5G
TCIAB3610-25GEA 2450 NOK
TCIAB3610-25G TCIAB3610-5G
5-Bromopyridine-2-carbonitrile ≥98.0% (by GC)
5-Bromopyridine-2-carbonitrile
Formel: C₆H₃BrN₂
MW: 183,01 g/mol
Kokepunkt: 110°C
MDL nummer: MFCD00234144
CAS-nummer: 97483-77-7
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72307892ac2d7a2ea02ca86aeff1c562262819c5&vid=72307892ac2d7a2ea02ca86aeff1c562262819c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR