N,N-Dimethylglycine ethyl ester ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Di-Me-Gly-OEt

D0726-25G D0726-500G
TCIAD0726-25GEA 485 NOK
TCIAD0726-25G TCIAD0726-500G
N,N-Dimethylglycine ethyl ester ≥98.0% (by GC)
N,N-Dimethylglycine ethyl ester
Formel: C₆H₁₃NO₂
MW: 131,17 g/mol
Kokepunkt: 40°C
Flash punkt: 32°C
MDL nummer: MFCD00009175
CAS-nummer: 33229-89-9
EINECS: 251-411-9
FN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d092648110886782d7ec0934654cd928136cb0b&vid=4d092648110886782d7ec0934654cd928136cb0b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR