3,4-Dihydro-2-methoxy-2H-pyran ≥97.0% (by GC)

Leverandør: TCI
Danger

D1554-25ML D1554-500ML
TCIAD1554-25MLEA 245 NOK
TCIAD1554-25ML TCIAD1554-500ML
3,4-Dihydro-2-methoxy-2H-pyran ≥97.0% (by GC)
3,4-Dihydro-2-methoxy-2H-pyran
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokepunkt: 127°C
Flash punkt: 16°C
MDL nummer: MFCD00006560
CAS-nummer: 4454-05-1
EINECS: 224-698-3
FN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=065ec5aa6ffe57e08d870fd1b49a9e8b42212f89&vid=065ec5aa6ffe57e08d870fd1b49a9e8b42212f89&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR