Benzoyl ferrocene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Benzoylferrocene

B0886-25G B0886-5G
TCIAB0886-25GEA 1725 NOK
TCIAB0886-25G TCIAB0886-5G
Benzoyl ferrocene ≥98.0% (by GC)
Benzoyl ferrocene
Formel: C₁₇H₁₄FeO
MW: 290,14 g/mol
MDL nummer: MFCD00001431
CAS-nummer: 1272-44-2
EINECS: 215-054-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08491c80d87208e25eec9eccc7673e20f960aef9&vid=08491c80d87208e25eec9eccc7673e20f960aef9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR