2-Bromo-1-indanol ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Bromo-1-hydroxyindane
B1300-25G
B1300-5G
TCIAB1300-25GEA
1125
NOK
TCIAB1300-25G
TCIAB1300-5G
2-Bromo-1-indanol ≥98.0% (by GC)
2-Bromo-1-indanol
Formel:
C₉H₉BrO MW: 213,07 g/mol Kokepunkt: 290 °C (1013 hPa) Smeltepunkt: 131…132 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00003798 CAS-nummer: 5400-80-6 EINECS: 226-442-6 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...