1-Indanon ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2,3-Dihydro-1H-inden-1-one

I0013-25G I0013-5G
TCIAI0013-25GEA 785 NOK
TCIAI0013-25G TCIAI0013-5G
1-Indanon ≥98.0% (by GC)
1-Indanon
Formel: C₉H₈O
MW: 132,16 g/mol
Kokepunkt: 245°C
Smeltepunkt: 37…40 °C
Tetthet: 1,1028 g/cm³ (40 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003785
CAS-nummer: 83-33-0
EINECS: 201-470-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9be235144e8d23dc201ea5c6dab45493310b1e6a&vid=9be235144e8d23dc201ea5c6dab45493310b1e6a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR