1-Bromo-2-methylnaphthalene ≥90.0% (by GC)

Leverandør: TCI
Warning

B1295-25G B1295-5G
TCIAB1295-25GEA 1025 NOK
TCIAB1295-25G TCIAB1295-5G
1-Bromo-2-methylnaphthalene ≥90.0% (by GC)
1-Bromo-2-methylnaphthalene
Formel: C₁₁H₉Br
MW: 221,1 g/mol
Kokepunkt: 157°C
Tetthet: 1,41 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003871
CAS-nummer: 2586-62-1
EINECS: 219-966-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=007397406858e22a7ca6d76c285a0462800a6378&vid=007397406858e22a7ca6d76c285a0462800a6378&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR