1,2-Dimetylnaftalen ≥95.0% (by GC)
Leverandør: TCI
D0747-1G
D0747-5G
TCIAD0747-1GEA
1475
NOK
TCIAD0747-1G
TCIAD0747-5G
1,2-Dimetylnaftalen ≥95.0% (by GC)
1,2-Dimetylnaftalen
Formel:
C₁₂H₁₂ MW: 156,23 g/mol Kokepunkt: 132°C Tetthet: 1,0219 g/cm³ (16.3 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004035 CAS-nummer: 573-98-8 EINECS: 209-364-7 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...