1,5-Dimetylnaftalen ≥97.0% (by GC)

Leverandør: TCI
Warning

D2764-1G D2764-5G
TCIAD2764-1GEA 865 NOK
TCIAD2764-1G TCIAD2764-5G
1,5-Dimetylnaftalen ≥97.0% (by GC)
1,5-Dimetylnaftalen
Formel: C₁₂H₁₂
MW: 156,23 g/mol
Kokepunkt: 265 - 266 °C (1013 hPa)
Smeltepunkt: 75 °C
MDL nummer: MFCD00004038
CAS-nummer: 571-61-9
EINECS: 209-338-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7eacfb34bfe0009e7a59f694363765cc343e606d&vid=7eacfb34bfe0009e7a59f694363765cc343e606d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR