2-Ethoxypyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
E0870-25G
E0870-5G
TCIAE0870-25GEA
1175
NOK
TCIAE0870-25G
TCIAE0870-5G
2-Ethoxypyridine ≥98.0% (by GC, titration analysis)
2-Ethoxypyridine
Formel:
C₇H₉NO MW: 123,15 g/mol Kokepunkt: 157°C Flash punkt: 52°C |
MDL nummer:
MFCD00023464 CAS-nummer: 14529-53-4 EINECS: 238-553-7 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...