2-brom-3-metylpyridin ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Bromo-3-picoline

B1894-25G B1894-5G
TCIAB1894-25GEA 1325 NOK
TCIAB1894-25G TCIAB1894-5G
2-brom-3-metylpyridin ≥97.0% (by GC)
2-brom-3-metylpyridin
Formel: C₆H₆BrN
MW: 172,02 g/mol
Kokepunkt: 219°C
MDL nummer: MFCD00239380
CAS-nummer: 3430-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fc5d4f3a8556aeef68fa1a69479ae67228b6623&vid=6fc5d4f3a8556aeef68fa1a69479ae67228b6623&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR