2,6-Lutidin ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,6-Dimethylpyridine

L0067-25ML L0067-500ML
TCIAL0067-25MLEA 380 NOK
TCIAL0067-25ML TCIAL0067-500ML
2,6-Lutidin ≥98.0% (by GC)
2,6-Lutidin
Formel: C₇H₉N
MW: 107,16 g/mol
Kokepunkt: 144°C
Smeltepunkt: –6 °C
Tetthet: 0,9252 g/cm³ (20 °C)
Flash punkt: 33°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006345
CAS-nummer: 108-48-5
EINECS: 203-587-3
FN: 1993
ADR: 3,III
Merck Indeks: 14,05616
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33463bc4e9570e7fefbdc83d49ebdfc4395e8196&vid=33463bc4e9570e7fefbdc83d49ebdfc4395e8196&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR