3,5-Lutidin ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3,5-Dimethylpyridine

L0068-25ML L0068-500ML
TCIAL0068-25MLEA 220 NOK
TCIAL0068-25ML TCIAL0068-500ML
3,5-Lutidin ≥98.0% (by GC)
3,5-Lutidin
Formel: C₇H₉N
MW: 107,16 g/mol
Kokepunkt: 172°C
Smeltepunkt: –9 °C
Tetthet: 0,94 g/cm³ (20 °C)
Flash punkt: 47°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006404
CAS-nummer: 591-22-0
EINECS: 209-708-6
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR