2-Adamantanol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol

A0720-25G A0720-5G
TCIAA0720-25GEA 1400 NOK
TCIAA0720-25G TCIAA0720-5G
2-Adamantanol ≥98.0% (by GC)
2-Adamantanol
Formel: C₁₀H₁₆O
MW: 152,24 g/mol
Smeltepunkt: 299…303 °C
Tetthet: 1,07 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00074744
CAS-nummer: 700-57-2
EINECS: 211-846-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b353b76d8fe856e16e484bc091a18c5688183a19&vid=b353b76d8fe856e16e484bc091a18c5688183a19&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR