Ethyl propionylacetate ≥97.0% (by GC)
Leverandør: TCI
Warning
Synonymer: Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate
K0031-25G K0031-5G
TCIAK0031-25GEA 1725 NOK
TCIAK0031-25G TCIAK0031-5G
Ethyl propionylacetate ≥97.0% (by GC)
Ethyl propionylacetate
Formel: C₇H₁₂O₃
MW: 144,17 g/mol
Kokepunkt: 191°C
Flash punkt: 77°C
MDL nummer: MFCD00009317
CAS-nummer: 4949-44-4
EINECS: 225-593-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5620c40816ecf30a2d43f8d312ef30233e8081f7&vid=5620c40816ecf30a2d43f8d312ef30233e8081f7&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %