Isobutyl propionate ≥98.0% (by GC)
Leverandør: TCI
Warning
Synonymer: Propionic acid Isobutyl ester
P0506-25ML P0506-500ML P0506-100ML
TCIAP0506-25MLEA 230 NOK
TCIAP0506-25ML TCIAP0506-500ML TCIAP0506-100ML
Isobutyl propionate ≥98.0% (by GC)
Isobutyl propionate
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokepunkt: 138°C
Flash punkt: 35°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009307
CAS-nummer: 540-42-1
EINECS: 208-746-0
FN: 2394
ADR: 3,III
Merck Indeks: 14,05150
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405ce34d96b9f84b1190e67fd335673e18a378c9&vid=405ce34d96b9f84b1190e67fd335673e18a378c9&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %