Dimethoxymethylvinylsilane ≥94.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Dimethoxy(methyl)(vinyl)silane

D3386-25G D3386-250G
TCIAD3386-25GEA 310 NOK
TCIAD3386-25G TCIAD3386-250G
Dimethoxymethylvinylsilane ≥94.0% (by GC)
Dimethoxymethylvinylsilane
Formel: C₅H₁₂O₂Si
MW: 132,23 g/mol
Kokepunkt: 104°C
Flash punkt: 3°C
MDL nummer: MFCD00129170
CAS-nummer: 16753-62-1
EINECS: 240-816-6
FN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d82b29e6e4456dc262472806a20e50dee5f1b4d&vid=6d82b29e6e4456dc262472806a20e50dee5f1b4d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR