4-Piperidinemethanol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: Piperidin-4-ylmethanol

P1653-25G P1653-5G
TCIAP1653-25GEA 2175 NOK
TCIAP1653-25G TCIAP1653-5G
4-Piperidinemethanol ≥97.0% (by GC, titration analysis)
4-Piperidinemethanol
Formel: C₆H₁₃NO
MW: 115,18 g/mol
Kokepunkt: 135°C
MDL nummer: MFCD00174228
CAS-nummer: 6457-49-4
FN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=344521350a3aea8c1b5ed3c6650b7ad75ce75129&vid=344521350a3aea8c1b5ed3c6650b7ad75ce75129&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 58.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR