1-Methyl-4-piperidinemethanol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

M2242-25G M2242-5G
TCIAM2242-25GEA 2350 NOK
TCIAM2242-25G TCIAM2242-5G
1-Methyl-4-piperidinemethanol ≥97.0% (by GC, titration analysis)
1-Methyl-4-piperidinemethanol
Formel: C₇H₁₅NO
Kokepunkt: 108°C
MDL nummer: MFCD03411903
CAS-nummer: 20691-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e36683170f35865373a13ac9bb5b21f4941cee1&vid=1e36683170f35865373a13ac9bb5b21f4941cee1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR