2,5-Dihydroxy-p-benzoquinone ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2,5-Dihydroxy-1,4-benzoquinone
D0574-25G
D0574-5G
TCIAD0574-25GEA
6050
NOK
TCIAD0574-25G
TCIAD0574-5G
2,5-Dihydroxy-p-benzoquinone ≥98.0% (by HPLC, titration analysis)
2,5-Dihydroxy-p-benzoquinone
Formel:
C₆H₄O₄ MW: 140,1 g/mol |
MDL nummer:
MFCD00001598 CAS-nummer: 615-94-1 EINECS: 210-454-3 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...