3,5-Di-tert-butyl-o-benzoquinone ≥98.0% (by GC)
Leverandør: TCI
D2430-25G
D2430-5G
TCIAD2430-25GEA
3000
NOK
TCIAD2430-25G
TCIAD2430-5G
3,5-Di-tert-butyl-o-benzoquinone ≥98.0% (by GC)
3,5-Di-tert-butyl-o-benzoquinone
Formel:
C₁₄H₂₀O₂ MW: 220,31 g/mol |
MDL nummer:
MFCD00001647 CAS-nummer: 3383-21-9 EINECS: 222-189-0 |
Spesifikasjon Testresultater
Appearance | Red to Dark blue to Black powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...