3-Methylbenzo[b]thiophene ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Methylbenzothiophene

M1952-1G M1952-5G
TCIAM1952-1GEA 580 NOK
TCIAM1952-1G TCIAM1952-5G
3-Methylbenzo[b]thiophene ≥97.0% (by GC)
3-Methylbenzo[b]thiophene
Formel: C₉H₈S
MW: 148,23 g/mol
Kokepunkt: 127°C
Smeltepunkt: –40°C
Flash punkt: 104°C
MDL nummer: MFCD00040243
CAS-nummer: 1455-18-1
EINECS: 215-934-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a88a972a86731d48d761039fb1d93b0e5bb0ea7&vid=2a88a972a86731d48d761039fb1d93b0e5bb0ea7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR