2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
5-Amino-1,3,4-thiadiazole-2-thiol
A0425-25G
A0425-500G
TCIAA0425-25GEA
270
NOK
TCIAA0425-25G
TCIAA0425-500G
2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel:
C₂H₃N₃S₂ MW: 133,2 g/mol Smeltepunkt: 234°C |
MDL nummer:
MFCD00003108 CAS-nummer: 2349-67-9 EINECS: 219-078-4 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in 1mol/L NaOH | faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...