3-Methylbutyl isovalerate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Isovaleric acid Isopentyl ester, Isovaleric acid Isoamyl ester, Isoamyl Isovalerate, Isopentyl 3-methylbutanoate, Isopentyl Isovalerate

I0196-25ML I0196-500ML
TCIAI0196-25MLEA 220 NOK
TCIAI0196-25ML TCIAI0196-500ML
3-Methylbutyl isovalerate ≥98.0% (by GC)
3-Methylbutyl isovalerate
Formel: C₁₀H₂₀O₂
MW: 172,27 g/mol
Flash punkt: 72°C
MDL nummer: MFCD00048369
CAS-nummer: 659-70-1
EINECS: 211-536-1
Merck Indeks: 14,05121
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=373f2faf0d4aad64dccf38b67f6c79cf8a6ae1b5&vid=373f2faf0d4aad64dccf38b67f6c79cf8a6ae1b5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR