6-Amino-1-tetralone 97%
Leverandør: Thermo Scientific
|
Danger
|
Synonymer:
6-Amino-3,4-dihydronaphthalen-1(2H)-one, 6-Amino-1,2,3,4-tetrahydronaphthalen-1-one
H32028.03
H32028.09
H32028.03EA
545
NOK
H32028.03
H32028.09
6-Amino-1-tetralone 97%
6-Amino-1-tetralone
Purity: 97%
CAS No.: 3470-53-9
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Melting Point: 128-130°
UN No.: 2811
MDL No.: MFCD00099462
Formel:
C₁₀H₁₁NO MW: 161,2 g/mol Smeltepunkt: 128…130 °C |
MDL nummer:
MFCD00099462 CAS-nummer: 3470-53-9 FN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...