5-Methoxy-2-tetralone ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 3,4-Dihydro-5-methoxy-2(1H)-naphthalenone

M1543-1G M1543-5G
TCIAM1543-1GEA 525 NOK
TCIAM1543-1G TCIAM1543-5G
5-Methoxy-2-tetralone ≥95.0% (by GC)
5-Methoxy-2-tetralone
Formel: C₁₁H₁₂O₂
MW: 176,22 g/mol
Kokepunkt: 165°C
Smeltepunkt: 37°C
MDL nummer: MFCD00064958
CAS-nummer: 32940-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feed35167ecc2c34eeef1221c6ea4b540143f7ed&vid=feed35167ecc2c34eeef1221c6ea4b540143f7ed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR