7-Methoxy-1-tetralone ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 7-Methoxy-3,4-dihydronaphthalen-1(2H)-one

M2092-25G M2092-5G
TCIAM2092-25GEA 1400 NOK
TCIAM2092-25G TCIAM2092-5G
7-Methoxy-1-tetralone ≥98.0% (by GC)
7-Methoxy-1-tetralone
Formel: C₁₁H₁₂O₂
MW: 176,22 g/mol
MDL nummer: MFCD00001696
CAS-nummer: 6836-19-7
EINECS: 229-916-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b8196bd8e51f73c10ee70f40d334daec9a9974a&vid=9b8196bd8e51f73c10ee70f40d334daec9a9974a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR