3-Methyl-2(5H)-furanone ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Hydroxy-2-methyl-2-butenoic acid gamma-Lactone

M1078-1ML M1078-25ML M1078-5ML
TCIAM1078-1MLEA 745 NOK
TCIAM1078-1ML TCIAM1078-25ML TCIAM1078-5ML
3-Methyl-2(5H)-furanone ≥97.0% (by GC)
3-Methyl-2(5H)-furanone
Formel: C₅H₆O₂
MW: 98,1 g/mol
Kokepunkt: 98°C
Flash punkt: 93°C
MDL nummer: MFCD00191545
CAS-nummer: 22122-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03ae7a709e0b72e0858f20c6bd3df83b6817dd0&vid=a03ae7a709e0b72e0858f20c6bd3df83b6817dd0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR