2-Pyrimidinecarbonitrile ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Cyanopyrimidine

C1518-1G C1518-25G C1518-5G
TCIAC1518-1GEA 340 NOK
TCIAC1518-1G TCIAC1518-25G TCIAC1518-5G
2-Pyrimidinecarbonitrile ≥98.0% (by GC)
2-Pyrimidinecarbonitrile
Formel: C₅H₃N₃
MW: 105,1 g/mol
MDL nummer: MFCD00160513
CAS-nummer: 14080-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c45c2a232dc7fe72adb4b9aaed88ffd2e7fdcba2&vid=c45c2a232dc7fe72adb4b9aaed88ffd2e7fdcba2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR