4,4''-Dibromo-p-terphenyl ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4,4''-Dibromo-1,1':4',1''-terphenyl

D3372-1G D3372-5G
TCIAD3372-1GEA 420 NOK
TCIAD3372-1G TCIAD3372-5G
4,4''-Dibromo-p-terphenyl ≥95.0% (by GC)
4,4''-Dibromo-p-terphenyl
Formel: C₁₈H₁₂Br₂
MW: 388,1000 g/mol
Smeltepunkt: 315°C
MDL nummer: MFCD00272889
CAS-nummer: 17788-94-2
FN: 3152
ADR: 9,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5843a99199de7713df47fd8146d7e7e29f4b51c&vid=d5843a99199de7713df47fd8146d7e7e29f4b51c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR