1,2,3,4-Tetrahydroisoquinoline ≥95.0% (by GC)

Leverandør: TCI
Warning

T1676-25G T1676-500G T1676-100G
TCIAT1676-25GEA 340 NOK
TCIAT1676-25G TCIAT1676-500G TCIAT1676-100G
1,2,3,4-Tetrahydroisoquinoline ≥95.0% (by GC)
1,2,3,4-Tetrahydroisoquinoline
Formel: C₉H₁₁N
MW: 133,19 g/mol
Kokepunkt: 233°C
MDL nummer: MFCD00006896
CAS-nummer: 91-21-4
EINECS: 202-050-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b83df6a46f967f99365659ad6fa8dbb7e9464557&vid=b83df6a46f967f99365659ad6fa8dbb7e9464557&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.0470 to 1.0500
Refractive index n20/D 1.5680 to 1.5710

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR