Benzyl carbamate ≥97.0% (by total nitrogen basis)
Leverandør: TCI
C0590-25G
C0590-250G
TCIAC0590-25GEA
485
NOK
TCIAC0590-25G
TCIAC0590-250G
Benzyl carbamate ≥97.0% (by total nitrogen basis)
Benzyl carbamate
Formel:
C₈H₉NO₂ MW: 151,17 g/mol |
MDL nummer:
MFCD00007965 CAS-nummer: 621-84-1 EINECS: 210-710-4 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 87.0 to 89.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...