Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride
G0102-500G G0102-25G
TCIAG0102-500GEA 810 NOK
TCIAG0102-500G TCIAG0102-25G
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99.0% (by titrimetric analysis)
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
Formel: C₄H₁₀ClNO₂
MW: 139,58 g/mol
MDL nummer: MFCD00012871
CAS-nummer: 623-33-6
EINECS: 210-787-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfec9ed1a0c82dd2c69ad32258c5754ddad19a77&vid=cfec9ed1a0c82dd2c69ad32258c5754ddad19a77&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 99.0 %
Water max. 1.0 %
Solubility in Water almost transparency