2-Aminoethyl hydrogen sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Sulfuric acid Mono(2-aminoethyl) Ester, 2-Aminoethyl Hydrogen sulphate

S0445-25G S0445-500G
TCIAS0445-25GEA 260 NOK
TCIAS0445-25G TCIAS0445-500G
2-Aminoethyl hydrogen sulphate ≥98.0% (by titrimetric analysis)
2-Aminoethyl hydrogen sulphate
Formel: C₂H₇NO₄S
MW: 141,1500 g/mol
Smeltepunkt: 279°C
MDL nummer: MFCD00008179
CAS-nummer: 926-39-6
EINECS: 213-135-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4021934fd115d60ae2e6ef8751580de9f11f2aa&vid=d4021934fd115d60ae2e6ef8751580de9f11f2aa&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR