1-Chloroisoquinoline ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C2097-25G
C2097-5G
TCIAC2097-25GEA
3475
NOK
TCIAC2097-25G
TCIAC2097-5G
1-Chloroisoquinoline ≥98.0% (by GC)
1-Chloroisoquinoline
Formel:
C₉H₆ClN MW: 163,61 g/mol Kokepunkt: 108°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00024134 CAS-nummer: 19493-44-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 33.0 to 38.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...