3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: DL-Lactide

L0091-25G L0091-100G
TCIAL0091-25GEA 970 NOK
TCIAL0091-25G TCIAL0091-100G
3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)
3,6-Dimethyl-1,4-dioxane-2,5-dione
Formel: C₆H₈O₄
MW: 144,13 g/mol
MDL nummer: MFCD00011685
CAS-nummer: 95-96-5
EINECS: 202-468-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1c2b03ef59ba4a7ff32780b53cb56302dffc81a&vid=c1c2b03ef59ba4a7ff32780b53cb56302dffc81a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR