2-Thienylacetyl chloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

T1751-250G T1751-25G
TCIAT1751-250GEA 5200 NOK
TCIAT1751-250G TCIAT1751-25G
2-Thienylacetyl chloride ≥98.0% (by GC, titration analysis)
2-Thienylacetyl chloride
Formel: C₆H₅ClOS
MW: 160.62 g/mol
Flash punkt: 85°C
MDL nummer: MFCD00005456
CAS-nummer: 39098-97-0
EINECS: 254-290-0
FN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83936551f8350ebc41c784aa7864d5befc91ac5&vid=d83936551f8350ebc41c784aa7864d5befc91ac5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Amber to Dark purple clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR