3-Amino-1H-1,2,4-triazole-5-thiol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol

A1455-25G A1455-250G A1455-5G
TCIAA1455-25GEA 340 NOK
TCIAA1455-25G TCIAA1455-250G TCIAA1455-5G
3-Amino-1H-1,2,4-triazole-5-thiol ≥98.0% (by HPLC, titration analysis)
3-Amino-1H-1,2,4-triazole-5-thiol
Formel: C₂H₄N₄S
MW: 116,15 g/mol
MDL nummer: MFCD00005231
CAS-nummer: 16691-43-3
EINECS: 240-735-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115cecce550a10cf1a2e20b39950349375edd2f4&vid=115cecce550a10cf1a2e20b39950349375edd2f4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR