5-Fluorouridine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione
F0636-1G
F0636-5G
TCIAF0636-1GEA
230
NOK
TCIAF0636-1G
TCIAF0636-5G
5-Fluorouridine ≥98.0% (by titrimetric analysis)
5-Fluorouridine
Formel:
C₉H₁₁FN₂O₆ MW: 262,19 g/mol Smeltepunkt: 185°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00036832 CAS-nummer: 316-46-1 EINECS: 206-260-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +15.0 to +20.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...